Learn More
United Scientific Supplies Class 3R Laser Pointers

Shop All United Scientific Supplies, Inc Products


Description
- Class 3R Laser Pointer with push-button switch
- Ideal for optical experiments
- Housed in rubberized aluminum
- Pen-style case with pocket clip
- Dimensions (L x D): 6 x 0.5 in.
- Maximum Output: 5 mW
- Wavelength: 532 nm
- Operates on (2) AAA batteries
- Available in Red, Green, Red/Green, and Violet
Specifications
Specifications
| Product Type | Laser Pointer, Class 3R |
| Description | Red/Green |
| For Use With (Application) | Optical Experiments |
| Length (English) | 6 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.