Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Laser Pointer with Holder
SDPSCI_ED

Catalog No. S89543 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89543 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89543 Supplier United Scientific™ Supplier No. UNLPH01A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This United Scientific™ Laser Pointer with Holder that allows you to securely hold a laser pointer at any angle and the clip can hold the toggle button in the on position for extended demonstrations and experiments.

This United Scientific Laser Pointer with Holder that allows you to securely hold a laser pointer at any angle. The clip can hold the toggle button in the on position for extended demonstrations and experiments.

  • The laser support rod is 8 in. long and has a vinyl coated clip to hold the laser pointer securely without scratching
  • The heavy cast iron base is 4 x 6 in. with black enamel finish
  • The 18 in. long zinc-plated steel rod has a diameter of 5/16 in.
  • The swivel clamp gives you the freedom to adjust the angle of the laser pointer 360°
  • The clamp is plated steel and accepts rods up to ¾ in. diameter (19 mm)
  • Length across clamps is 4.5 in.
  • The red laser pointer operates on a push-button switch and are housed in a rubberized aluminum pen-style case
  • The laser is Class 3R and is 6 in. long x 0.5 in. diameter
  • Red laser pointer has a maximum output of 5 mW and a wavelength of 650 nm (± 10 nm)
  • Range of beam is 100 meters
  • Expected operating life is 1000 hours
  • Operates on two AAA batteries (which are included).

Specifications

Grade 5 to 12
Format Kit
Product Type Laser Pointer Holder
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.