Learn More
Sigma Aldrich Fine Chemicals Biosciences Laser Tough-SpotsR diam. 3
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7435843840EA
Encompass
Tough-Spots are chemically inert polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths conventional and cryogenic freezer temperatures most organic solvents caustic agents and other challenges without peeling becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Catalog No. 50-290-0528
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.