Learn More
Description
- Blank, printable polyester labels for Laser Printers
- CSA Accepted material when printed with designated laser printer toners
Specifications
Specifications
| Description | Chemical, high temperature, humidity, smear, UV and weather-Resistant |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | Yellow |
| Finish | Matte |
| Format | Label |
| Material | Polyester (B-747) |
| Printable Sides | Single-side Printable |
| Print Technology | Laser |
| Product Type | Laser Printable Label |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.