Learn More
Invitrogen™ LAT Monoclonal Antibody (LAT-01)

Description
This antibody reacts with an intracellular epitope of LAT, a 36-38 kDa transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. This antibody will not cross-react with mouse.
Specifications
Specifications
| Antigen | LAT |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | LAT-01 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | LAT |
| Gene Accession No. | O43561 |
| Gene Alias | 36 kDa phospho-tyrosine adapter protein; 36 kDa phospho-tyrosine adaptor protein; Lat; LAT1; linker for activation of T cells; linker for activation of T cells, transmembrane adaptor; linker for activation of T-cells; linker for activation of T-cells family member 1; linker protein; p36-38; pp36 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.