Learn More
MilliporeSigma™ Supelco™ Latch-Lid™ TLC Developing Chambers
Catalog No. 111016103 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
10 x 10 cm Plates
20 x 20 cm Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-6103 10 x 10 cm Plates
11-100-9742 20 x 20 cm Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6103 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2660191EA
Add to Cart
Add to Cart
Latch-Lid™ units have a unique stainless steel latching device that holds the lid and tank firmly in place.
Latch-Lid™ units have a unique stainless steel latching device that holds the lid and tank firmly in place. This permits complete saturation of the chamber with solvent vapor for uniform development of chromatograms. Glass tank features flat ground tops, inner and outer rims beveled for safety, and flat ground bottoms for stability. Accessorize with multi-plate TLC racks for simultaneous development of up to six TLC plates. Racks not included.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) TLC
For Use With (Equipment) 10 x 10 cm Plates
Includes Racks not included.
Product Line Latch-Lid
Latch-Lid is a trademark of General Glassblowing, Inc.