Learn More
MSA™ Latchways™ Vertical Ladder Lifeline Kit Attachment Device
Catalog No. 19141466 Shop All MSA Products
Change view
Click to view available options
Product Type: LadderLatch attachments device
Catalog No.
19-141-466 Product Type: LadderLatch attachments device
1 options
Catalog No. 19-141-466 Supplier MSA™ Supplier No. 3104L00
Add to Cart
Add to Cart
Designed to connect the user, wearing a full body harness, to a Latchways™ cable system. MSA™ Latchways™ Vertical Ladder Lifeline Kit Attachment Device allows hands-free operation for system users.
  • For use with MSA™ Latchways™ Vertical Ladder Lifeline Kits
  • Unique starwheel allows the device to pass over cable supports without the need to detach
  • Provides an easy-to-install solution when fall protection is necessary
Specifications
Product Type LadderLatch Attachments Device
Quantity 1 Each
Certifications/Compliance ANSI Z359.16 – 2016, CSA Z59.2.5, OSHA 1910 Walking-Working Surfaces ladder fall protection, EN 353:1, AS/NZ 1891.4, AOCS01-00-SCP-Z06-Compiled MSA Self Cert Form - Z359.16 VLL
Description The Latchways LadderLatch device is the only approved attachment device for use with MSA Latchways Vertical Ladder Kits.