Learn More
Cayman Chemical Latent TGF-1humnrecombI 500ug
Supplier: Cayman Chemical 30606500 UG
Source: Recombinant human C-terminal His-tagged TGF-β1 expressed in HEK293 cells • Amino acids: 1-390 (full length) • MW: 42.4 kDa for LAP-bound TGF-β1
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.