Learn More
Novus Biologicals™ LATEX 400nm Black Mini Antibody Conjugation Kit
Shop All Novus Biologicals Products

Description
The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.
Kit Contents:
- 1 vial of 10x Quencher buffer
- 1 vial of Resuspension buffer
- 1 vial of Reaction buffer B
- 1 vial of Reaction buffer A
- 4 or 10 Mini vials of 400nm Black LATEX™
Specifications
Specifications
| Target | LATEX™ 400nm Black Mini |
| Content And Storage | -20°C |
| Quantity | 40 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.