Learn More
PIP™ Latex Boots
Catalog No. 17010554 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-554 X-Large
17-010-555 2X-Large
17-010-553 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-010-554 Supplier PIP™ Supplier No. 8400/XL
Add to Cart
Add to Cart
Designed to wear over shoes or boots in manufacturing and/or environmental clean up.
  • 'Nuke Boot' style
  • Heavy weight high density rubber shoe cover
  • Textured sole for traction
  • Lightweight and easy to store
  • 100% waterproof
Specifications
Color Yellow
Material Latex
Size X-Large
Toe Material Rubber
Disposable Reusable
Sterility Non-sterile
Waterproof Water Repellent
Product Type Overboot