Learn More
Description
The kit utilizes a simple spin column to clean-up the antibody by buffer exchanging to remove the unwanted buffer components. The kit can also be used for concentrating your antibody by recovering the antibody in a smaller volume than the volume initially added to the Spin Cartridge.
Kit Contents:
- 1 vial 20x Reaction buffer A
- 1 vial 20x Reaction buffer B
- 4 spin cartridge / collecting tube assemblies
Specifications
Specifications
| Target | LATEX™ and Europium Conjugated |
| Content And Storage | 4°C |
| Product Line | Novus |
| Quantity | 4 Units |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
