Learn More
Columbia Cleanroom Latex - Free, Cleanroom Notebook
Description
- Printed and manufactured in Portland, OR, USA. College rule, engineering grid, or a combination of both.
- 100 pages (50 sheets).
- 22 pound blue cleanroom paper, latex-free.
- Translucent, heavy-duty polyethylene cover (made in the USA).
- USA-made poly coil.
- Handmade in the USA.
- Class 10 or ISO 3 certified.
- Side Spiral with 3 x 5 in., 5.5 x 8.5 in., 8.5 x 11 in., A4, A5 (European)
- 30 notebooks per case.
Specifications
Specifications
| Material | Paper |
| Ruled | College Ruled |
| Color | White, Blue |
| Dimensions (L x W) | 8.5 x 11 in. |
| Product Type | Notebook |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.