Learn More
MOLECULAR DEPOT LLC Lp-PLA2 mAb - 1 mg
Supplier:  MOLECULAR DEPOT LLC A2010035
Encompass_Preferred
Lp-PLA2 Monoclonal Antibody (1 mg) is an affinity-purified mouse monoclonal antibody specific to human Lipoprotein-associated phospholipase A2 (Lp-PLA2) a blood-circulating phospholipase that is involved in the metabolism of pro-inflammation Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more Several tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only
Catalog No. 50-000-13480
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.