Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Lattice Clamp

Catalog No. 04999123 Shop All Eisco Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04-999-123 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-999-123 Supplier Eisco™ Supplier No. CH0664C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For lattice building and rod coupling.

  • Uses 12.5 mm dia. rods
  • Fitted with screws for Allen wrench

Specifications

Type Lattice Clamp
Material Metal
For Use With (Equipment) Lattice building and rod coupling
For Use With (Application) Lattice building and rod coupling
Color Silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.