Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Lattice Lab System, Set No. 3
SDPSCI_ED

Catalog No. S43732 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43732 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43732 Supplier United Scientific Supplies Supplier No. LLS003
Add to Cart
Add to Cart

This lattice lab system features an assortment of aluminum rods, including ten rods that are 48 in. long.

  • This lattice lab system features an assortment of aluminum rods, including ten rods that are 48 in. long.
  • Assembly only requires a screwdriver.
  • Foot plates and hook connectors are made of cast alloy while the support rods are made of hard aluminum.
  • Foot plate, 2-5/16 in. diameter 6/Ea.
  • Set screw for foot plate 6/Ea.
  • Anchoring screw for foot plate 18/Ea.
  • Hook connector with thumbscrew 42/Ea.
  • Support rod, 2 in. long, 1/2 in. diameter 10/Ea.
  • Support rod, 18 in. long, 1/2 in. diameter 3/Ea.
  • Support rod, 36 in. long, 1/2 in. diameter 2/Ea.
  • Support rod, 48 in. long, 1/2 in. diameter 10/Ea.

Specifications

Grade 9 to 12
Format Kit
Product Type Lattice Lab System, Support Stands
Languages English
Subject Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.