Learn More
Lauda Brinkmann Lp LAUDA ECO E 20G
Supplier: Lauda Brinkmann Lp L001200
Recirculating water bath, LAUDA ECO E 20G, heating and circulating bath (20L) for temperature control of both internal and external applications. Temp range of 20 to 200 degreesC. 0.55 Bar Pump. 1.3 kW Heater 115V;60Hz This item is non returnable*
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.