Learn More
Lauda Brinkmann Lp LAUDA ECO ET 6G
Supplier:  Lauda Brinkmann Lp L001207
Encompass_Preferred
Recirculating water bath, LAUDA ECO ET 6G, heating and circulating clearview bath (6L) for temperature control of both internal and external applications. Temp range of 20 to 100 degreesC. 0.55 Bar Pump. 1.3 kW Heater 115V;60Hz This item is non returnable*
Catalog No. 50-305-651
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.