Learn More
Sigma Organic Chemistry lauric acid diethanolam | 1G | MFCD00045982
Supplier: Sigma Organic Chemistry CDS0105881G

lauric acid diethanolam , 1G
About this Item:
MDL #: MFCD00045982
Molecular Weight: 287.44
UNSPSC Code: 12352200
Chemical Formula: C16H33NO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.