Learn More
Medchemexpress LLC 1-propanaminium, 3-carboxy-N,N,N-tri(methyl-d3)-2-[(1-oxododecyl)oxy]-, chloride (1:1), (2R)- | 2245711-25-3 | 98.0% | C19H29D9ClNO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY130321S110MG
Encompass_Preferred
Lauroyl-L-carnitine-d9 chloride is the deuterium-labeled (d9) chloride salt of lauroyl-L-carnitine supplied as an isotope-labeled reference standard for analytical and research use. It is intended for use as an internal standard or tracer in mass spectrometry, quantitative assays, and metabolic or pharmacokinetic studies, providing a labeled analog of the unlabeled compound.
  • Deuterium-labeled (d9) isotope suitable for use as an internal standard in mass spectrometry.
  • Chloride salt form to match common analytical reference materials.
  • High purity (98.0%) for reliable analytical performance.
  • Provided in small, reference quantities tailored for analytical workflows.
  • Stable when stored sealed under refrigerated conditions (≈2-8°C).
Catalog No. 50-002-60025
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.