Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lauryl Tryptose Broth (Dehydrated)

Catalog No. R660514
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R660514 2.5 kg
OXCM0451B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R660514 Supplier Thermo Scientific™ Supplier No. CM0451R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect coliform organisms in water, waste water and dairy products with this selective medium.

Detect coliform organisms in water, waste water and dairy products with selective Thermo Scientific™ Oxoid™ Lauryl Tryptose Broth (Dehydrated), while conforming to the formula detailed by American Public Health Association. Lauryl Tryptose Broth is designed to promote a rich growth and copious gas production from small inocula of coliform organisms.

Use Lauryl Tryptose Broth for detection of coliform organisms in water, wastewater and dairy products for the presence of indicator groups such as coliforms. Besides safety, indicator groups provide a measure of food quality and spoilage potential. One of the most common groups of bacteria employed as indicator organisms by the food industry are the coliforms, which can be regarded as a subgroup within the Enterobacteriaceae

  • Selective: sodium lauryl sulphate selects for coliform group
  • Recommended by APHA1 for the Most Portable Number (MPN) presumptive test of coliforms in water, effluent or sewage.

The advantage of using Lauryl Tryptose Broth is that, in addition to giving the fermentation reaction typical of MacConkey Broth, it can also be tested for the presence of indole.

The APHA1 recommend that Lauryl Tryptose Broth should be used for the Most Probable Number (MPN) presumptive test of coliforms in waters, effluent or sewage as a confirmatory test of lactose fermentation with gas production for milk samples (APHA2) and for the detection of coliforms in foods (APHA3 ).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Lauryl Tryptose Broth (Lauryl Sulphate Broth)
Form Powder
Media Type Lauryl Tryptose Broth
Product Type Nutrient Broth
Quantity 2.5 kg
Yield For 70.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.