Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Layilin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114008 Supplier Invitrogen™ Supplier No. PA5114008
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CYKVIYFHDT SRRLNFEEAK EACRRDGGQL VSIESEDEQK LIEKFIENLL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 91%; Dog: 92%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Rabbit: 93%; Rat: 86%; Mouse: 79%;.

Layilin is a cell surface receptor encoded by the LAYN gene, known for its involvement in cell adhesion and motility processes. It is a member of the C-type lectin-like domain protein family and plays a crucial role in mediating interactions with the extracellular matrix. Layilin is associated with cytoskeletal reorganization, impacting cellular processes such as shape change, migration, and stabilization of cell-matrix contacts. It has been shown to interact with other proteins like talin, contributing to the linkage between the cell membrane and the actin cytoskeleton, which is vital for maintaining cell structure and facilitating movement. Layilin's role in cell adhesion and motility makes it significant in developmental processes and wound healing. Additionally, dysregulation of layilin expression has been implicated in cancer metastasis, where altered adhesive properties and increased motility contribute to tumor progression and dissemination. Research is ongoing to explore the detailed mechanisms of layilin's function and its potential as a target for therapeutic intervention in diseases characterized by abnormal cell migration and adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Layilin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LAYN
Gene Accession No. Q6UX15
Gene Alias E030012M19Rik; FLJ30977; FLJ31092; Gm511; layilin; LAYN; LYAN; RGD1564444; UNQ208/PRO234
Gene Symbols LAYN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human LAYN (aa 49-98).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143903
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.