Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB-Agar, Lennox, Large Capsules

Catalog No. MP113002326 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity: 454g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP113002326 Quantity: 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP113002326 Supplier MP Biomedicals™ Supplier No. 113002326
Add to Cart
Add to Cart
This item is not returnable. View return policy

Culture media used for the growth of bacteria.

  • Typically contain 10g of tryptone and 5g of yeast extract per liter, and vary in salt
TRUSTED_SUSTAINABILITY

Specifications

Broth Type LB Broth
Format Large capsules
Media Type Bacterial media
Product Type LB Agar
Quantity 454 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.