Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LB agar, Miller Microbiologically Tested, MilliporeSigma™ Supelco™

Catalog No. 111004054 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4054 1 kg
11-100-4668 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4054 Supplier MilliporeSigma Supelco Supplier No. L30271KG
Add to Cart
Add to Cart

Contains: tryptone, yeast extract, sodium chloride and agar

TRUSTED_SUSTAINABILITY

Specifications

Description Contains: tryptone, yeast extract, sodium chloride and agar
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.