Learn More
Teknova LB Agar Plates with Animal Free Soytone and Spectinomycin-50. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Supplier:  Teknova L15748PK
Encompass_Preferred
1.69% Animal-Free Soytone, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar,50µg/mL Spectinomycin Dihydrochloride Pentahydrate
Catalog No. 50-238-7538
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.