Learn More
Teknova LB Agar Plates with Apramycin-100, 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Supplier:  Teknova L10508PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Apramycin
Catalog No. 50-238-7718