Learn More
Teknova LB Agar Plates with Gentamicin-30, 100mm, 20 plates/sleeve, Sterile. 8 pack.
SDP
Supplier:  Teknova L10548PK
Encompass_Preferred
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,30µg/mL Gentamicin Sulfate
Catalog No. 50-238-7719
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.