Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ LB Broth, Miller (Granulated)

Catalog No. BP97232 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2000 g
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP9723-2 2000 g Poly Bottle
BP97235 5 kg Poly Pail
BP9723-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP9723-2
  • $770.50
    $654.65 / Each of 1

    Save $115.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge

missing translation for 'validMainframeMsg'

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 25g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Format: Granulated powder
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type LB Broth
Product Type Bacterial Culture
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) Microbiology Media
Packaging Type Poly Bottle
Quantity 2000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.