Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ LB Broth, Miller (Pre-Buffered Capsules)

Catalog No. BP97315 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 mL Capsules, 5 kg
25 x 25 g
500 x 1 g
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP97315 1000 mL Capsules, 5 kg Poly Pail
BP9731-500 25 x 25 g Poly Bottle
BP9732-500 500 x 1 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP97315
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 1 capsule per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
  • Product is in capsule form
  • Each capsule contains 25g of broth
  • Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
  • pH will vary with temperature
TRUSTED_SUSTAINABILITY

Specifications

Format Pre-Buffered Capsules
Media Type LB Broth
Product Type Microbiology Media
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, TRIS/TRIS HCl 1.5g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Pail
Quantity 1000 mL Capsules, 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.