Learn More
United States Biological Corporation LB Broth Miller, Animal Free (Powder) (Miller's LB broth, Luria-Bertani broth), 500g
Catalog No. NC1616741 Shop All United States Biological Corporation Products
Encompass_Preferred
LB Broth media formulations have been industry standards for the cultivation of Escherichia coli since the 1950’s (1,2,3,4). These media have been widely used in molecular microbiology applications for the preparation of plasmid DNA and recombinant proteins (1,5). The media are nutrient-rich formulations which provide peptides and peptones, vitamins, and trace elements.
Catalog No. NC1616741 Supplier United States Biological Corporation Supplier No. L152001500G
LB Broth media formulations have been industry standards for the cultivation of Escherichia coli since the 1950’s (1,2,3,4). These media have been widely used in molecular microbiology applications for the preparation of plasmid DNA and recombinant proteins (1,5). The media are nutrient-rich formulations which provide peptides and peptones, vitamins, and trace elements.