Learn More
Kd Medical Inc LB KAN 25MG/ML
Supplier: Kd Medical Inc BPL2578
LB Kanamycin plates. Used for the maintenance and propagation of E. coli, selective for kanamycin resistance. Ten 100mm plates per sleeve. Concentration of Kanamycin is 25mg/ml.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.