Learn More
Teknova LB Luria Agar Plates with 10mM Magnesium Sulfate. 86x128mm, 30mL fill, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Supplier:  Teknova L21334PK
Encompass_Preferred
1.00% Tryptone, 0.50% Yeast Extract,0.05% Sodium Chloride, 1.50% Agar,10mM Magnesium Sulfate Heptahydrate
Catalog No. 50-238-7754
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.