Learn More
Invitrogen™ LBP Monoclonal Antibody (RR433-8)

Description
Endotoxin level < 24 EU/mg. This antibody reacts highly specific with mouse natural and recombinant LBP. The antibody is a type II antibody blocking the LPS binding to CD14. The MA1-40030 immunogen is mouse natural and recombinant LBP. MA1-40030 detects LBP from mouse samples. MA1-40030 has been successfully used in ELISA, Functional Assays and Western blot applications.
Specifications
Specifications
| Antigen | LBP |
| Applications | ELISA, Functional Assay, Western Blot |
| Classification | Monoclonal |
| Clone | RR433-8 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene | LBP |
| Gene Accession No. | Q61805 |
| Gene Alias | acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; Bpifd2; hypothetical protein LOC512242; LBP; lipopolysaccharide binding protein; lipopolysaccharide-binding protein; lipopolysaccharide-binding protein LPSBP; LPS-binding protein; LPSBP; Ly88 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.