Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560662 Supplier Thermo Scientific Supplier No. PA560662
Only null left
Add to Cart
Add to Cart

LBX2 Polyclonal Antibody

LBX2 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen LBX2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LBX2
Gene Accession No. Q6XYB7
Gene Alias lady bird-like homeobox 2; lady bird-like homeobox 2 homolog; ladybird homeobox 2; ladybird homeobox homolog 2; ladybird homeobox homolog 2 (Drosophila); ladybird homeobox protein homolog 2; ladybird-like homeobox 2; ladybird-like homeodomain protein 2; LBX2; Lbx2h; LP3727; RGD1561172; Transcription factor LBX2
Gene Symbols LBX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LBX2. Recombinant protein control fragment (Product #RP-99931).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.