Learn More
QA Bio Inc QA-Bio, Inc. • LudgerSep R2 4.6x150mm HPLC Column for Glycan Analysis
Supplier:  QA Bio Inc LSUR22.1X100
Encompass
R2 4.6x150mm HPLC Column for Glycan Analysis • The LudgerSep R2 4.6x150mm column has been developed for HPLC analysis of 2-AA labeled monosaccharides labeled with 2-AA using the LudgerTag Monosaccharide Analysis Kit. • Dimensions • 4.6 x 150mm
Catalog No. 50-355-844
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.