Learn More
MilliporeSigma™ Supelco™ LC-MS Post column Flow Splitters, Fixed
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Description | Split Ratio = 10:1 |
| For Use With (Application) | Liquid Chromatography |