Learn More
MBL International Corporation Anti-LC3 pAb (Polyclonal Antibody)
SDP
Supplier:  MBL International Corporation PM036
Encompass_Preferred
Clonality:Polyclonal; Application:FCM|ICC|IHC|IP|WB; Conjugate:Unlabeled; Species Reactivity:Hamster|Human|Mouse|Rat; Size:100 µl; Target:LC3; This LC3 antibody is validated for multiple applications (WB, IHC, ICC and IP) and isoform detection. PM036 has more than 1,000 citations where it used to generate data showing autophagy in research papers.  This is a polyclonal antibody that is raised in rabbit and is reactive with human, hamster, mouse, rat. First LC3 antibody to market. Numerous publications available. LC3 Polyclonal Antibody.
Catalog No. 50-507-41
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.