Learn More
MBL International Corporation Anti-LC3 (Human) mAb (Monoclonal Antibody)
SDP
Supplier:  MBL International Corporation M1523
Encompass_Preferred
Clonality:Monoclonal; Application:FCM|ICC|IH|Immuno-EM|IP|WB; Conjugate:Unlabeled; Species Reactivity:Hamster|Human|Mouse|Rat; Size:200 µg/100 µL; Target:LC3; This LC3 antibody is validated for multiple applications (WB, ICC, IH, IP, FCM, Immuno-EM) and has numerous citations. This is a monoclonal antibody targeting LC3 for FCM, ICC, IP, WB.More information about LC3 and autophagy can be found in our Blog. In addition you may download our Autophagy Pathway Poster.LC3 Monoclonal Antibody.
Catalog No. 50-505-77
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.