Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LC3-II Enrichment Kit (Western Blot)

Catalog No. 1710232 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-102-32 15 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-32 Supplier MilliporeSigma™ Supplier No. 1710232

Includes

LC3-II Enrichment Component Box

  • Autophagy Reagent A: One vial, lyophilized
  • 10x Enrichment Buffer: One bottle containing 10mL 10x Enrichment Buffer
  • 5x Assay Buffer: One bottle containing 15mL 5x Assay Buffer
  • 10x RIPA Lysis Buffer: One bottle containing 1mL 10x RIPA Lysis Buffer

Western Blot Detection Component Box

  • Protease Inhibitor Cocktail: One vial containing 20 μL of 1000x Protease Inhibitor Cocktail
  • Anti-LC3 antibody: One vial containing 20μL of 1000x anti-LC3 rabbit monoclonal antibody
  • Anti-TOMM22 antibody: One vial containing 20μL of 1000x anti-TOMM22 mouse monoclonal antibody
  • Goat anti-Mouse IgG HRP, Secondary Antibody: One vial containing 30μL of 2000X goat anti-mouse HRP conjugate
  • Goat anti-Rabbit IgG HRP, Secondary Antibody: One vial containing 30μL of 2000X goat anti-rabbit HRP conjugate

Requires

  • Pipettors and tips capable of accurately measuring 1-1000μL
  • Graduated serological pipettes
  • Microcentrifuge tubes
  • Mechanical vortex
  • Rocker
  • Distilled or deionized water
  • 1x Dulbecco's Phosphate Buffered Saline (D-PBS)
  • 1x Tris-buffered saline containing 0.05% Tween-20 (TBST)
  • 3% Non-fat dry milk diluted in TBST (TBST-MILK)
  • Earle's Balanced Salt Solution (EBSS)

Enables sensitive and accurate quantification of autophagosome density by utilizing selective permeabilization procedure that removes cytosolic LC3-I and retains autophagosome-bound LC3-II

  • Enables sensitive and accurate quantification of autophagosome density by utilizing a selective permeabilization procedure that removes cytosolic LC3-I and retains autophagosome-bound LC3-II
  • Reveals quantities of LC3-II, without interference from LC3-I, by Western blotting analysis
  • The kit also includes a lysosomal inhibitor, to prevent degradation of LC3-II that occurs upon fusion of the autophagosome with the lysosome
  • An antibody against a mitochondrial outer membrane protein TOMM22 is included for use as a control for specificity of permeabilization, to ensure that organelles remain intact after treatment
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The kit may be used for up to 4 months after receipt. Avoid repeated freeze/thaw cycles, and aliquot if necessary
For Use With (Application) Western Blotting
Quantity 15 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.