Learn More
Description
This antibody detects both LC3A and LC3B. Western blot bands are seen at ~19 kDa, representing LC3-I, and ~17 kDa, representing LC3-II. The observed molecular weight of the protein may vary from the listed predicted molecular weight due to post translational modifications, post translation cleavages, relative charges, and other experimental factors. Recommended positive controls: LC3B overexpression lysate, mouse brain whole tissue lysate (adult whole normal), human brain whole tissue lysate (adult whole normal), Neuro2a whole cell lysate, Neuro2a Chloroquine-treated/untreated cell lysate, HeLa chloroquine-treated/untreated cell lysate This antibody is predicted to react with Bovine based on 100% sequence homology.
Specifications
Specifications
| Antigen | LC3A/LC3B |
| Applications | ChIP Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MAP1LC3B2 |
| Gene Accession No. | A6NCE7, O41515, Q2HJ23, Q62625, Q6XVN8, Q91VR7, Q9CQV6, Q9GZQ8, Q9H492 |
| Gene Alias | 1010001C15Rik; 1010001H21Rik; 4922501H04Rik; Atg8; ATG8E; ATG8F; Autophagy-related protein LC3 A; autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 A; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3a; LC3b; MAP1 light chain 3-like protein 1; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 A; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 A; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; Map1lc3; Map1lc3a; MAP1LC3B; MAP1LC3B-a; microtubule associated protein 1 light chain 3 alpha; microtubule associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 alpha; Microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; Microtubule-associated proteins 1A/1B light chain 3B; Mpl3; wu:fb60g11; zbs559; zgc:56434; zgc:77094 |
| Gene Symbols | MAP1LC3A, MAP1LC3B, MAP1LC3B2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
