Learn More
TARGETMOL CHEMICALS INC LCB 03-0110 DIHYDROCHLOR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T32613
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LCB 03-0110 Dihydrochloride is a potent inhibitor of the c-Src kinase (IC50 = 1.3 nM) and tyrosine kinases in the BTK and SYK family as well as in the DDR2 family. It inhibits LPS-induced activation of macrophages and TGF-(beta)1-induced activation of fibroblasts in vitro inhibits activation of macrophages and fibroblasts and inhibits scarring in wound healing models. purity: 99%
Catalog No. 50-258-0325
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.