Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCE1A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7210180 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4642 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4642 Supplier Proteintech Group Inc Supplier No. 249561AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LCE1A belongs to the late cornified envelope (LCE) gene cluster within the epidermal differentiation complex (EDC) on chromosome 1. The LCE cluster contains multiple conserved genes that encode stratum corneum proteins, and these genes are expressed relatively late during fetal assembly of the skin cornified envelope.

Specifications

Antigen LCE1A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene LCE1A
Gene Accession No. Q5T7P2
Gene Alias late cornified envelope 1A, Late envelope protein 1, LCE1A, LEP1
Gene Symbols LCE1A
Host Species Rabbit
Immunogen LCE1A Fusion Protein Ag21047
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353131
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.