Learn More
Abcam LCL161, 10MG
Supplier:  Abcam AB27361610MG
Encompass_Preferred
MW 500.6 Da, Purity >99%. Small molecule SMAC mimetic inhibitor of multiple IAP (Inhibitor of apoptosis) family proteins. (IC₅₀ = 10.23 μM). Inhibits cell proliferation and viability in two human hepatocellular carcinoma cells (Hep3B, IC₅₀ = 10, and PLC5, IC₅₀ = 19).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2917
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.