Learn More
Apexbio Technology LLC LCL161, 100mg. Cas: 1005342-46-0 MFCD: MFCD23160049. Antagonist of IAPs inhibitor
SDP
Supplier:  Apexbio Technology LLC A3541100
Encompass_Preferred
LCL161 is a small molecular antagonist of the inhibitor of apoptosis (IAP) with IC50 value of 10.23 M in Hep3B cells [1].IAP is a family of proteins which are firstly found in virus and to inhibit the apoptosis of the infected hosts. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4811
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.