Learn More
Medchemexpress LLC HY-16582 100mg , LDE225 (Diphosphate) NVP-LDE 225 Diphosphate;Erismodegib Diphosphate CAS:1218778-77-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY16582100MG
Encompass_Preferred
Medchemexpress, HY-16582 100mg LDE225 (Diphosphate) NVP-LDE 225 Diphosphate;Erismodegib Diphosphate CAS:1218778-77-8 Formula:C26H32F3N3O11P2 IC50: 1.3 nM (mSmo), 2.5 nM (hSmo) Purity:98% LDE225 Diphosphate is a potent and selective Smo antagonist with IC 50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3142
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.