Learn More
Bioassay Systems QuantiChrom™ LDH Cytotoxicity Assay Kit. For rapid quantitative determination of cytotoxicity based on lactate dehydrogenase released into cell culture medium.
SDP
Catalog No. 501078328 Shop All Bioassay Systems Products
Encompass_Preferred
For rapid quantitative determination of cytotoxicity based on lactate dehydrogenase released into cell culture medium. Evaluation of toxic compounds, toxins, detergents, environmental pollutants and physical trea™ent on cell lysis. Key Features: Safe. Non-radioactive assay (cf. chromium release assay). Fast. The whole procedure take 20 min. Robust and amenable to HTS. Single reagent, "mix-incubate-measure" type assay. High-throughput assay in 96-well plates allows simultaneous processing tens of thousands of samples per day. Method: OD500nm. Samples: Cell culture. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests.
Catalog No. 50-107-8328 Supplier Bioassay Systems Supplier No. C2LD100
For rapid quantitative determination of cytotoxicity based on lactate dehydrogenase released into cell culture medium. Evaluation of toxic compounds, toxins, detergents, environmental pollutants and physical trea™ent on cell lysis. Key Features: Safe. Non-radioactive assay (cf. chromium release assay). Fast. The whole procedure take 20 min. Robust and amenable to HTS. Single reagent, "mix-incubate-measure" type assay. High-throughput assay in 96-well plates allows simultaneous processing tens of thousands of samples per day. Method: OD500nm. Samples: Cell culture. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests.