Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHA Recombinant Rabbit Monoclonal Antibody (4H18L6), Alexa Fluor™ Plus 647

Catalog No. 702747RP647
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-274-7RP647 50 μL
50-274-5776 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-274-7RP647 Supplier Invitrogen™ Supplier No. 702747RP647
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This 702747RP647 antibody (4H18L6) has been tested by western blot and immunocytochemistry analysis of PC-3 cells. This may be used for immunocytochemistry at 5 μg/mL and for western blotting at less than or equal to 1.25 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 658 nm; Emission: 675 nm; Laser: Red Laser Filtration: 0.2 μm post-manufacturing filtered.

Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers. Mutations in the LDHA gene can lead to Glycogen storage disease 11 (GSD11).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHA
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4H18L6
Concentration 1.0 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8
Gene LDHA
Gene Accession No. P00338
Gene Alias Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; GSD11; HEL-S-133P; l7R2; Lactate Dehydrogenase; lactate dehydrogenase 1, A chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase A; lactate dehydrogenase A4; lactate dehydrogenase A-like; lactate dehydrogenase C; lactate dehydrogenase C variant 3; lactate dehydrogenase M; lactate dehydrogenase-A; lactate dehydrogenase-M; lactate dehydrogenase-M (EC 1.1.1.27); Lactic Dehydrogenase; LDH; LDH muscle subunit; LDH testis subunit; Ldh1; Ldh-1; Ldh3; Ldha; LDH-A; ldha protein; ldha1; LDH-A4 muscle-specific isozyme; glycolytic enzyme; Ldhc; LDH-C; Ldhm; LDH-M; LDH-X; L-lactate dehydrogenase; L-lactate dehydrogenase A chain; L-lactate dehydrogenase C chain; M-LDH; OTTHUMP00000165206; PIG19; proliferation-inducing gene 19; renal carcinoma antigen NY-REN-59
Gene Symbols LDHA
Host Species Rabbit
Immunogen Peptides corresponding to human LDHA [1) aa211-aa229 2) aa2-aa19].
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3939
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.