Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHAL6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590143 Supplier Invitrogen™ Supplier No. PA590143
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSWTVPVVRA SQRVSSVGAN FLCLGMALCP RQATRIPLNG TWLFTPVSKM ATVKSELIER FTSEKPVHHS KVSIIGTGSV; Positive Samples: Mouse testis, Rat kidney.

LDHAL6B belongs to the LDH/MDH superfamily. LDH family. The specific function of the protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHAL6B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LDHAL6B
Gene Alias 4933402O15Rik; AI326310; lactate dehydrogenase A like 6B; lactate dehydrogenase A-like 6; lactate dehydrogenase A-like 6B; LDH6B; Ldhal; LDHAL6; Ldhal6b; LDHL; L-lactate dehydrogenase A-like 6B; testicular tissue protein Li 105
Gene Symbols LDHAL6B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human LDHAL6B (NP_1499721).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106557, 369018
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.