Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDHC Recombinant Rabbit Monoclonal Antibody (K01_1T33)

Catalog No. MA565132
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA565132 50 μL
MA563673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA565132 Supplier Invitrogen™ Supplier No. MA565132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

LDHC Recombinant Monoclonal Antibody for Western Blot

Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHC
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene LDHC
Gene Accession No. P07864
Gene Alias Cancer/testis antigen 32; CT32; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase C; lactate dehydrogenase c variant 1; lactate dehydrogenase c variant 3; lactate dehydrogenase c variant 4; lactate dehydrogenase c variant 5; lactate dehydrogenase C4; lactate dehydrogenase-C; lactate dehydrogenase-X; LDH testis subunit; Ldh3; Ldh-3; LDHC; LDH-C; Ldhc4; LDH-C4; LDHX; Ldh-x; L-lactate dehydrogenase C; L-lactate dehydrogenase C chain; MGC111073; MGC127715 protein; testis secretory sperm-binding protein Li 234P
Gene Symbols LDHC
Host Species Rabbit
Immunogen A synthetic peptide of human Lactate Dehydrogenase C.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3948
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.