Learn More
Apexbio Technology LLC LDK378 dihydrochloride 1380575-43-8 5mg
SDP
Supplier:  Apexbio Technology LLC A35445
Encompass_Preferred
LDK378 dihydrochloride (CAS 1380575-43-8) is a potent inhibitor of anaplastic lymphoma kinase (ALK) a receptor tyrosine kinase in the insulin receptor superfamily Demonstrating an in vitro IC50 of 200 pM against ALK LDK378 also inhibits IGF-1R InsR and STK22D with IC50 values of 8 nM 7 nM and 23 nM respectively In cellular assays LDK378 suppresses proliferation of Ba/F3 cells expressing NPM-ALK and Karpas-299 human non-Hodgkin lymphoma cells with IC50 values of 22 8 nM and 26 nM This compound serves as a valuable tool for investigating ALK-driven signaling pathways and potential therapeutic strategies in oncology research
Catalog No. 50-224-0726
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.