Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ldl Uptake Assay Kit, Flow Cytometry, Adherent Cells, 48 Tests

Catalog No. 502586454
Encompass_Preferred
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6454 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6454 Supplier Abcam Supplier No. AB2362081X48TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for the analysis of LDL uptake in adherent cell samples via flow cytometry.

  • Detection method: Flow cytometry-fluorescent
  • Sample types: Adherent cells
  • Cited in: > 3 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule LDL
For Use With (Equipment) Flow Cytometer
No. of Reactions 48 Tests
Detection Location In-Cell Detection
Format Kit
Detection Method Fluorescence
Sample Type Adherent cells
Quantity 48 Tests
For Use With (Application) LDL uptake assay, Flow cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.